Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dopamine D1R/DRD1 Antibody (SG2-D1a), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. N1106017A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1106017A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1106017A32 Supplier Novus Biologicals Supplier No. NB11060017AF532

Mouse Monoclonal Antibody

Dopamine D1R/DRD1 Monoclonal specifically detects Dopamine D1R/DRD1 in Mouse, Rat, Bovine (Negative), Canine (Negative), Human (Negative), Porcine (Negative), Rabbit (Negative), Sheep (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating.

Specifications

Antigen Dopamine D1R/DRD1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone SG2-D1a
Conjugate Alexa Fluor 532
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias D(1A) dopamine receptor, D1DR, DADR, Dopamine D1 receptor, dopamine receptor D1, DRD1A
Gene Symbols DRD1
Host Species Mouse
Immunogen Recombinant rat Dopamine Receptor D1 protein near the C-terminus. [Swiss-ProtNo. P18901]
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 1812
Test Specificity This antibody is specific for Dopamine Receptor D1A. There is no cross-reactivity with D1B (D5) receptor.
Target Species Mouse, Rat, Bovine, Canine, Human, Pig, Rabbit, Sheep
Content And Storage Store at 4°C in the dark.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.