Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dopamine D2R/DRD2 Antibody (HL1478) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB295174 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB295174 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB295174 Supplier Novus Biologicals Supplier No. NBP325443
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Dopamine D2R/DRD2 Monoclonal antibody specifically detects Dopamine D2R/DRD2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Functional Assay Assay

Specifications

Antigen Dopamine D2R/DRD2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Functional Assay
Classification Monoclonal
Clone HL1478
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:10000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin, Functional Assay
Formulation PBS
Gene Alias D(2) dopamine receptor, D2DR, D2R, Dopamine D2 receptor, dopamine receptor D2, dopamine receptor D2 isoform, seven transmembrane helix receptor
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-term region of Human Dopamine D2R/DRD2. The exact sequence is proprietary.
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline GPCR, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 1813
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.