Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dopamine D2R/DRD2 Antibody, Novus Biologicals™
SDP

Catalog No. NB036587 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB036587 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB036587 Supplier Novus Biologicals Supplier No. NBP288996
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Dopamine D2R/DRD2 Polyclonal antibody specifically detects Dopamine D2R/DRD2 in Human, Mouse, Rat, Avian samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen Dopamine D2R/DRD2
Applications Western Blot, ELISA, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS containing 50% glycerol
Gene Alias D(2) dopamine receptor, D2DR, D2R, Dopamine D2 receptor, dopamine receptor D2, dopamine receptor D2 isoform, seven transmembrane helix receptor
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the sequence near the N-terminus of mouse Dopamine D2R/DRD2.
Molecular Weight of Antigen 51 kDa
Purification Method Peptide affinity purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline GPCR, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 1813
Target Species Human, Mouse, Rat, Avian
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.