Learn More
Description
Specifications
Specifications
| Antigen | Dopamine D3R/DRD3 |
| Applications | Western Blot, ELISA, Immunofluorescence, Immunoprecipitation, Dot Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000-1:15000, ELISA 1:50000, Immunocytochemistry/Immunofluorescence 5 μg /ul, Immunoprecipitation, Dot Blot 1:50000 |
| Formulation | Contains Tris, HCl/glycine buffer (pH 7.4-7.8), 30% glycerol, 0.5% BSA, along with cryo-protective agents, and HEPES (exact storage and stabilization buffer information is proprietary) with 0.02% Sodium Azide |
| Gene Alias | D(3) dopamine receptor, D3DR, Dopamine D3 receptor, dopamine receptor D3, essential tremor 1, ETM1, FET1, MGC149204, MGC149205 |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide corresponding to unique amino acid sequence on Dopamine Receptor D3 protein, peptide was covalently modified. Sequence: CQACHVSPELYRATTWGY |
| Purification Method | Affinity-purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
