Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dopamine D5R/DRD5 Antibody (889022) [Janelia Fluor™ 646], Novus Biologicals™
SDP

Catalog No. NB373245 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB373245 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB373245 Supplier Novus Biologicals Supplier No. FAB82861J
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Dopamine D5R/DRD5 Monoclonal specifically detects Dopamine D5R/DRD5 in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen Dopamine D5R/DRD5
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 889022
Conjugate Janelia Fluor 646
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias D(1B) dopamine receptor, D(5) dopamine receptor, D1beta dopamine receptor, Dopamine D5 receptor, dopamine receptor D1B, dopamine receptor D5, DRD1BDBDR, DRD1L2MGC10601
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human Dopamine D5 R/DRD5, Met1-His477, Accession # P21918
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, GPCR, Neuroscience, Neurotransmission, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1816
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.