Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dopamine D5R/DRD5 Antibody (SG4-D1b), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. N1106019A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1106019A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1106019A94 Supplier Novus Biologicals Supplier No. NB11060019AF594

Mouse Monoclonal Antibody

Dopamine D5R/DRD5 Monoclonal specifically detects Dopamine D5R/DRD5 in Mouse, Rat, Human (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Dopamine D5R/DRD5
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone SG4-D1b
Conjugate Alexa Fluor 594
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias D(1B) dopamine receptor, D(5) dopamine receptor, D1beta dopamine receptor, Dopamine D5 receptor, dopamine receptor D1B, dopamine receptor D5, DRD1BDBDR, DRD1L2MGC10601
Gene Symbols DRD5
Host Species Mouse
Immunogen Recombinant rat Dopamine Receptor D5 (within the last 118 residues of the C-terminus). [Swiss-ProtNo. P25115]
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 1816
Target Species Mouse, Rat, Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.