Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dopamine D5R/DRD5 Antibody (SG4-D1b), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB11060019U Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11060019U 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11060019U Supplier Novus Biologicals Supplier No. NB11060019UV

Mouse Monoclonal Antibody

Dopamine D5R/DRD5 Monoclonal specifically detects Dopamine D5R/DRD5 in Mouse, Rat, Human (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Dopamine D5R/DRD5
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone SG4-D1b
Conjugate DyLight 350
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Gene Alias D(1B) dopamine receptor, D(5) dopamine receptor, D1beta dopamine receptor, Dopamine D5 receptor, dopamine receptor D1B, dopamine receptor D5, DRD1BDBDR, DRD1L2MGC10601
Gene Symbols DRD5
Host Species Mouse
Immunogen Recombinant rat Dopamine Receptor D5 (within the last 118 residues of the C-terminus). [Swiss-Prot# P25115]
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, GPCR, Neuroscience, Neurotransmission, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1816
Test Specificity Rat. Reactivity with mouse has had mixed results. It does not react with human protein.
Target Species Mouse, Rat, Human (Negative)
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.