Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Doppel Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB315636 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB315636 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB315636 Supplier Novus Biologicals Supplier No. NBP297004MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Doppel Polyclonal antibody specifically detects Doppel in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)

Specifications

Antigen Doppel
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution ELISA, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias dJ1068H6.4, DOPPEL, DPLprion gene complex, downstream, MGC41841, Prion protein 2, prion protein 2 (dublet), PrPLPprion-like protein doppel
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human Doppel (Accession#: NP_036541.2; Met1-Gly152)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neurodegeneration, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 23627
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.