Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Dorsomorphin dihydrochloride
GSA_VAGREENER_CHOICE

Catalog No. 309310
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-931-0 10 mg
30-935-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 30-931-0 Supplier Tocris Bioscience™ Supplier No. 3093/10
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Potent AMPK inhibitor

Dorsomorphin dihydrochloride is a potent inhibitor of AMP-activated protein kinase (AMPK) (Ki = 109 nM). Displays no significant activity on several structurally related kinases including ZAPK, SYK, PKCθ, PKA and JAK3. Inhibits AMPK activation induced by AICAR (Cat. No. 2840) and Metformin (Cat. No. 2864). Dorsomorphin dihydrochloride also inhibits bone morphogenetic protein (BMP) type I receptors (ALK2, ALK3 and ALK6), promotes cardiomyogenesis in mouse embryonic stem cells (ESCs) in vitro and promotes neural differentiation of hPSCs as part of a chemical cocktail. Shown to induce autophagy in cancer cell lines via a mechanism independent of AMPK inhibition.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 1219168-18-9
Molecular Formula C24H27Cl2N5O

Specifications

Quantity 10 mg
Formula Weight 472.41
Purity >98%
Molecular Weight (g/mol) 472.414
Chemical Name or Material Dorsomorphin dihydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.