Learn More
BRANDTECH™ BRAND™ Dosing Element for Dispensette™ S Trace Analysis Bottletop Dispensers
Click to view available options
For Use With (Equipment):
Dispensette S Trace Analysis
Description
- Replacement dosing element for BRAND™ Dispensette™ S Trace Analysis 1 to 10 mL
- Fits instruments with tantalum or platinum-iridium spring
- Includes certificate of performance
Specifications
Specifications
Volume (Metric) | 10 mL |
For Use With (Equipment) | Dispensette S Trace Analysis |
Includes | Quality Certificate |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov