Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DOT1L Antibody (OTI1D8), PerCP, Novus Biologicals™
SDP

Catalog No. NB288268 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB288268 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB288268 Supplier Novus Biologicals Supplier No. NBP272297PCP

Mouse Monoclonal Antibody

DOT1L Monoclonal antibody specifically detects DOT1L in Human samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen DOT1L
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1D8
Conjugate PerCP
Dilution Western Blot, Immunohistochemistry
Formulation PBS
Gene Alias DOT1-like protein, DOT1-like, histone H3 methyltransferase (S. cerevisiae), EC 2.1.1.43, H3-K79-HMTase, KIAA1814DKFZp586P1823, KMT4H3 lysine-79 specific, Lysine N-methyltransferase 4
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-434 of human DOT1L (NP_115871) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chromatin Research, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 84444
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.