Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DOT1L Antibody, Novus Biologicals™
SDP

Catalog No. NB10040845 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10040845 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10040845 Supplier Novus Biologicals Supplier No. NB10040845
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 11 publications

DOT1L Polyclonal specifically detects DOT1L in Human, Mouse samples. It is validated for Chemotaxis, ChIP Assay, ChIP Cytometry, ChIP sequencing (ChIP-seq), IF/IHC, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown, Western Blot.

Specifications

Antigen DOT1L
Applications Chemotaxis, ChIP Assay, ChIP sequencing (ChIP-seq), Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunohistochemistry 1:200-1:1000, Immunoprecipitation 2-5 ug/mg lysate, Immunohistochemistry-Paraffin 1:200-1:1000, Chromatin Immunoprecipitation (ChIP), Chromatin Immunoprecipitation Sequencing, Knockdown Validated
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. Q8TEK3
Gene Alias DOT1-like protein, DOT1-like, histone H3 methyltransferase (S. cerevisiae), EC 2.1.1.43, H3-K79-HMTase, KIAA1814DKFZp586P1823, KMT4H3 lysine-79 specific, Lysine N-methyltransferase 4
Gene Symbols DOT1L
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue1000 and 1050 of human DOT1-Like using the numbering given in entry NP_115871.1 (GeneID 84444).
Molecular Weight of Antigen 185 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84444
Test Specificity Mouse reactivity reported in scientific literature (PMID: 23861670). Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Rhesus Monkey, White-tufted-ear marmoset and Northern white-cheeked gibbon.
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.