Learn More
Poltex Double Bulk Bin, 8 in. Wide Comp
Description
Simple two-compartment bulk bin with two compartments, open top for filling and front for dispensing.
Durable and cleanable PETG construction.
Choose from:
- Wall or countertop mount with rubber bumpons on base
- VHB tape mount (very high bond tape included)
- Magnet mount (powerful magnets included)
Specifications
Specifications
| Type | Dispensing Bin |
| Material | PETG |
| Color | Clear |
| Shape | Rectangular |
| Mounting Type | Magnetic Mount |
| Autoclavable | Not Autoclavable |