Learn More
PIP™ Double-Layer Full-Face Nomex™ Hood
PIP™ Double-Layer Full-Face Nomex™ Hood are flame resistant balaclavas that offer limited chemical splash protection along with excellent wear and durability
$99.00 - $117.00
Specifications
Product Type Fire Protectant Hood
Size Universal
Material Nomex
Disposable Reusable
Quantity 1 Ea.
View More Specs
Products 3
Catalog Number Mfr. No. Color Design Type Price Quantity & Availability  
Catalog Number Mfr. No. Color Design Type Price Quantity & Availability  
19-100-450
View Documents
PIP™
906100NOM7
White Tri-Cut
Each for $112.00
Only null left
Add to Cart
19-100-451
View Documents
PIP™
906100NOM7B
White Straight Cut
Each for $99.00
Only null left
Add to Cart
19-100-452
View Documents
PIP™
906100NOM7BLKB
Black Straight Cut
Each for $117.00
Only null left
Add to Cart
Description
Description
A key factor in the thermal protection Nomex™ provides is that it carbonizes and thickens when exposed to the intense heat of fires. This increases the protective barrier between the heat source and the skin and minimizes burn injury. This tough, protective barrier stays supple and flexible until it cools, which gives valuable extra seconds of protection when it is most needed. 
  • Protective qualities will not wash out
  • Fabric meets OSHA requirements for flame resistance
  • TPP Rating 20-23
  • Meets NFPA 1971 requirements
  • One Size Fits Most
Specifications
Specifications
Fire Protectant Hood
Nomex
1 Ea.
2-ply head and bib
Universal
Reusable
Hood Only
Heat/Fire Applications
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.