Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Doublecortin Antibody (3E1), FITC, Novus Biologicals™
SDP

Catalog No. NBP192684F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP192684F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP192684F Supplier Novus Biologicals Supplier No. NBP192684F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Doublecortin Monoclonal specifically detects Doublecortin in Human, Mouse, Rat, Porcine, Bovine, Equine samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Doublecortin
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 30
Concentration 1mg/mL
Conjugate FITC
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Gene Accession No. O43602
Gene Alias doublecortin, Doublin, FLJ51296, Lissencephalin-X, Lis-X, X-linked (doublecortin)
Gene Symbols DCX
Host Species Mouse
Immunogen Recombinant full length human Lis-A isoform of Doublecortin purified from E. coli. [UniProt# O43602]
Molecular Weight of Antigen 49.3 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1641
Test Specificity Human, Mouse, Rat, Cow, Pig and other Mammals.
Target Species Human, Mouse, Rat, Pig, Bovine, Equine
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.