Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Doublecortin Antibody, Novus Biologicals™
SDP

Catalog No. NBP269992 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP269992 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP269992 Supplier Novus Biologicals Supplier No. NBP269992
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Doublecortin Polyclonal specifically detects Doublecortin in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Peptide ELISA.

Specifications

Antigen Doublecortin
Applications Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Flow Cytometry 10 ug/mL, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 10 ug/mL, Immunohistochemistry-Paraffin 6 ug/mL, Peptide ELISA 1:6000
Formulation TBS (pH 7.3) with 0.5% BSA with 0.02% Sodium Azide
Gene Alias doublecortin, Doublin, FLJ51296, Lissencephalin-X, Lis-X, X-linked (doublecortin)
Gene Symbols DCX
Host Species Goat
Immunogen Peptide with sequence DRSHCQSLRFHQN, from the internal region of the protein sequence according to NP_000546.2.
Purification Method Antigen Affinity-purified
Quantity 100 μg
Research Discipline Neuroscience, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1641
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.