Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Doublecortin Monoclonal Antibody (01), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA543039
This item is not returnable.
View return policy
Description
Specificity: Human Doublecortin/DCX
In the developing cortex, cortical neurons must migrate over long distances to reach the site of their final differentiation. DCX is a cytoplasmic protein which appears to direct neuronal migration by regulating the organization and stability of microtubules. It contains two doublecortin domains, which bind microtubules. In addition, this protein interacts with LIS1, the regulatory gamma subunit of platelet activating factor acetylhydrolase, and this interaction is important to proper microtubule function in the developing cortex. Mutations in the gene encoding DCX are a cause of X-linked lissencephaly.
Specifications
Doublecortin | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
O43602 | |
DCX | |
Recombinant Human Doublecortin/DCX Protein. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Immunocytochemistry | |
01 | |
Unconjugated | |
DCX | |
DBCN; Dbct; DC; Dcn; DCX; doublecortex; doublecortin; doublin; lissencephalin-X; lissencephaly, X-linked (doublecortin); LISX; Lis-X; LIS-XD; Neuronal migration protein doublecortin; OTTHUMP00000023860; OTTHUMP00000216316; SCLH; XLIS | |
Mouse | |
Protein A | |
RUO | |
1641 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction