Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Doublecortin Monoclonal Antibody (3E1)
Mouse Monoclonal Antibody
$526.00 - $1800.00
Specifications
Antigen | Doublecortin |
---|---|
Clone | 3E1 |
Concentration | 1 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA547314
|
Invitrogen™
MA547314 |
500 μL |
Each for $1,800.00
|
|
|||||
PIMA547412
|
Invitrogen™
MA547412 |
100 μL |
Each for $526.00
|
|
|||||
Description
Doublecortin Monoclonal Antibody for Western Blot, ICC/IF, IHC
In the developing cortex, cortical neurons must migrate over long distances to reach the site of their final differentiation. DCX is a cytoplasmic protein which appears to direct neuronal migration by regulating the organization and stability of microtubules. It contains two doublecortin domains, which bind microtubules. In addition, this protein interacts with LIS1, the regulatory gamma subunit of platelet activating factor acetylhydrolase, and this interaction is important to proper microtubule function in the developing cortex. Mutations in the gene encoding DCX are a cause of X-linked lissencephaly.Specifications
Doublecortin | |
1 mg/mL | |
Immunohistochemistry, Western Blot, Immunocytochemistry | |
Unconjugated | |
Mouse | |
RUO | |
PBS with 50% glycerol and 5mM sodium azide | |
O43602, O88809, Q9ESI7 | |
13193, 1641, 84394 | |
Recombinant full length human Lis-A isoform of doublecortin expressed in and purified from E. coli. | |
Antibody |
3E1 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Monoclonal | |
Liquid | |
IgG2a | |
Human, Mouse, Rat | |
DCX | |
DBCN; Dbct; DC; Dcn; DCX; doublecortex; doublecortin; doublin; lissencephalin-X; lissencephaly, X-linked (doublecortin); LISX; Lis-X; LIS-XD; Neuronal migration protein doublecortin; OTTHUMP00000023860; OTTHUMP00000216316; SCLH; XLIS | |
DCX | |
Primary | |
Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title