Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Doxorubicin hydrochloride
GREENER_CHOICESDP

Catalog No. 225250 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-525-0 50 mg
22-521-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-525-0 Supplier R&D Systems™ Supplier No. 2252/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Antitumor antibiotic agent. Inhibits DNA topoisomerase II

Doxorubicin hydrochloride is a 14-hydroxylated version of Daunorubicin (Cat. No. 1467) that is naturally fluorescent. Doxorubicin is an antitumor antibiotic agent that inhibits DNA topoisomerase II. It is a DNA intercalator that inhibits nucleic acid synthesis and induces apoptosis. Doxorubicin reduces intracellular tau levels. Doxorubicin also promotes formation of free radicals for the disruption of membrane lipids and DNA strands. Doxorubicin fluorescence is quenched upon intercalation into the DNA; while binding to histones or partitioning into the phospholipid phase of PEG-phospholipid micelles or hydrophobic cores of polymeric micelles, increases Doxorubicin fluorescence. Doxorubicin fluorescence enables the monitoring of the localization of the drug within lipid bilayers and liposomal delivery systems and interaction of the drug with DNA and other macromolecules, as well as drug efflux pump activities.

Chemical Identifiers

CAS 25316-40-9
Molecular Formula C27H30ClNO11

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 579.99
Purity >98%
Molecular Weight (g/mol) 579.98
Chemical Name or Material Doxorubicin hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.