Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Doxorubicin Hydrochloride Inhibitor

Low molecular weight, organic compound that may help regulate biological process.

Specifications

CAS 25316-40-9
Formula Weight 579.99g/mol
For Use With (Application) Inhibits apoptosis
Molecular Formula C30H41FN4O12
Solubility Soluble to 50nM in DMSO, 50nM water
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB225210MG View Documents Novus Biologicals™
NB225210MG
10mg This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
NB225250MG View Documents Novus Biologicals™
NB225250MG
50mg This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Biological Activity: Antitumor antibiotic agent that inhibits DNA topoisomerase II. DNA intercalator that inhibits nucleic acid synthesis and induces apoptosis. Reduces intracellular tau levels.

Storage: Store at 4°C.

Note: Inhibitors are guaranteed for 1 year from date of receipt.

Specifications

Specifications

25316-40-9
Inhibits apoptosis
Soluble to 50nM in DMSO, 50nM water
Doxorubicin hydrochloride
579.99g/mol
C30H41FN4O12
Store at 4°C
Orange

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit