Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Doxorubicin HydroChloride, PROMO, Thermo Scientific Chemicals

Catalog No. J64000MFPM
Click to view available options
Quantity:
50 mg

Doxorubicin hydrochloride is the hydrochloride salt of doxorubicin, isolated from the bacterium Streptomyces peucetius, and is considered to be an anthracycline antibiotic with antineoplastic activity.

General Description

  • Doxorubicin hydrochloride, isolated from the bacterium Streptomyces peucetius, is an anthracycline antibiotic with antineoplastic activity and is related to daunorubicin
  • It can intercalate between base pairs in the DNA helix preventing DNA replication
  • It also inhibits the nucleic acid synthesis and can induce apoptosis
  • It can inhibit DNA topoisomerase II, resulting in an increased and stabilized cleavable enzyme-DNA linked complex during DNA replication, and prevents nucleotide strand ligation after double-strand breakage
  • Doxorubicin hydrochloride forms oxygen free radicals that result in cytotoxicity and lead to the lipid peroxidation of cell membrane lipids

Application

  • This compound displays a fluorescent property allowing the measurement of drug efflux pump activity and examining the role of subcellular organelles (Golgi complex and lysosome) in the sequestration of drugs
  • It has been used in cell viability assays
  • It is used as a drug in polymeric PLGA-based microparticulate drug delivery and to develop in vitro doxorubicin-resistant HepG2 cells (HepG2-DR) and K562 cells (K562-DR)
  • It is a substrate of the MRP1 transporter
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25316-40-9
Molecular Formula C27H30ClNO11
Molecular Weight (g/mol) 579.98 g/mol
MDL Number MFCD00077757,MFCD00077757,MFCD00941448
InChI Key MWWSFMDVAYGXBV-FGBJBKNOSA-N
Synonym doxorubicin hydrochloride, adriamycin, doxorubicin hcl, adriacin, adriblastina, rubex, adriamycin hydrochloride, adriblastin, adriamycin pfs
PubChem CID 129626538
IUPAC Name hydrogen (8S,10S)-10-[(4-amino-5-hydroxy-6-methyloxan-2-yl)oxy]-6,8,11-trihydroxy-8-(2-hydroxyacetyl)-1-methoxy-5,7,8,9,10,12-hexahydrotetracene-5,12-dione chloride
SMILES [H+].[Cl-].COC1=CC=CC2=C1C(=O)C1=C(O)C3=C(C[C@](O)(C[C@@H]3OC3CC(N)C(O)C(C)O3)C(=O)CO)C(O)=C1C2=O

Specifications

Melting Point 216°C (decomposition)
Color Orange-Red
Quantity 50 mg
Beilstein 4229251
Merck Index 14,3439
Solubility Information Soluble in water.
Physical Form Solid
Chemical Name or Material Doxorubicin hydrochloride
EINECSNumber 246-818-3
RTECSNumber QI9295900
TSCA No

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.