Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPAGT1 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017234 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017234 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB017234 Supplier Novus Biologicals Supplier No. NBP281923
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DPAGT1 Polyclonal specifically detects DPAGT1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen DPAGT1
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, ELISA, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias ALG7, CDG1J, CDG-Ij, D11S366, DGPTDPAGT2UAGT, dolichyl-phosphate (UDP-N-acetylglucosamine)N-acetylglucosaminephosphotransferase 1 (GlcNAc-1-P tra, dolichyl-phosphate (UDP-N-acetylglucosamine)N-acetylglucosaminephosphotransferase 1 (GlcNAc-1-P transferase), dolichyl-phosphate alpha-N-acetylglucosaminyltransferase, EC 2.7.8, EC 2.7.8.15, G1PT, GlcNAc-1-P transferase, GPTDPAGT, N-acetylglucosamine-1-phosphate transferase, UDP-GlcNAc:dolichyl-phosphate N-acetylglucosaminephosphotransferase, UDP-N-acetylglucosamine--dolichyl-phosphateN-acetylglucosaminephosphotransferase, UGAT
Gene Symbols DPAGT1
Host Species Rabbit
Immunogen DPAGT1 antibody was raised against a 17 amino acid synthetic peptide near the amino terminus of human DPAGT1. The immunogen is located within amino acids 30 - 80 of DPAGT1. Amino Acid Squence: QDLNKTSRQQIPESQGV
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1798
Test Specificity At least four isoforms of DPAGT1 are known to exist; this antibody will recognize the two longest isoforms. DPAGT1 antibody is predicted to not cross-react with UHRF1BP1.
Target Species Human, Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.