Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ DPCPX
GREENER_CHOICESDP

Catalog No. 0439100 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-391-00 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 04-391-00 Supplier Tocris Bioscience™ Supplier No. 0439/100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective A1 antagonist

Potent and selective A1 adenosine receptor antagonist, both in vitro and in vivo. Ki values are 3.9, 130, 50 and 4000 nM for human A1, A2A, A2B and A3 receptors respectively.

Chemical Identifiers

CAS 102146-07-6
Molecular Formula C16H24N4O2

Specifications

Quantity 100 mg
Formula Weight 304.39
Purity >99%
Molecular Weight (g/mol) 304.39
Chemical Name or Material DPCPX
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.