Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPCR-1 Antibody (213201) [Allophycocyanin], Novus Biologicals™
SDP

Catalog No. NB372470 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB372470 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB372470 Supplier Novus Biologicals Supplier No. FAB3347A
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DPCR-1 Monoclonal specifically detects DPCR-1 in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen DPCR-1
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 213201
Conjugate APC
Dilution Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias bCX105N19.6, C6orf37, diffuse panbronchiolitis critical region 1, MGC126710, MGC126712, PBLTdiffuse panbronchiolitis critical region 1 protein
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human DPCR-1, Met1-Arg235, Accession # AAH69477
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 135656
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.