Learn More
Invitrogen™ DPF1 Polyclonal Antibody

Description
Immunogen sequence: VLEALLCAET GEKKIELKEE ETIMDCQKQQ LLEFPHDLEV EDLEDDIPRR KNRAKGKAYG IGGLRKRQDT ASLEDR Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
Specifications
Specifications
| Antigen | DPF1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | DPF1 |
| Gene Accession No. | Q92782 |
| Gene Alias | Baf45b; BRG1-associated factor 45B; D4, zinc and double PHD fingers family 1; double PHD fingers 1; DPF1; NEU BAF45b; NEUD4; neuro-d4; neuro-d4 homolog; neuronal d4 domain family member; zinc finger protein neuro-d4 |
| Gene Symbols | DPF1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.