Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPF2 Antibody, DyLight 680, Novus Biologicals™
SDP

Catalog No. NB103326 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB103326 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB103326 Supplier Novus Biologicals Supplier No. NBP297764FR
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DPF2 Polyclonal specifically detects DPF2 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen DPF2
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 680
Formulation 50 mM Sodium Borate
Gene Alias Apoptosis response zinc finger protein, D4, zinc and double PHD fingers family 2BAF45D, MGC10180, Protein requiem, REQBRG1-associated factor 45D, requiem, apoptosis response zinc finger, ubi-d4, UBID4requiem, apoptosis response zinc finger gene, zinc finger protein ubi-d4
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human DPF2 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 5977
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.