Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPH2 Antibody (OTI3A12) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP270587 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270587 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270587 Supplier Novus Biologicals Supplier No. NBP270587
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DPH2 Monoclonal specifically detects DPH2 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, CyTOF-ready.

Specifications

Antigen DPH2
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, CyTOF
Classification Monoclonal
Clone OTI3A12
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:2000, Flow Cytometry 1:100, Immunohistochemistry 1:150, Immunocytochemistry/Immunofluorescence 1:100, CyTOF-ready
Formulation Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative
Gene Alias diphthamide biosynthesis protein 2, Diphthamide biosynthesis protein 2 homolog-like 2, diptheria toxin resistance protein required for diphthamide biosynthesis-like 2, diptheria toxin resistance protein required for diphthamide biosynthesis-like 2(S. cerevisiae), DPH2 homolog, DPH2 homolog (S. cerevisiae), DPH2L2diphthamide biosynthesis-like protein 2, DPH2-like 2, DPH2-like 2 (S. cerevisiae), HsDph2
Gene Symbols DPH2
Host Species Mouse
Immunogen Full length human recombinant protein of human DPH2(NP_001375) produced in HEK293T cell.
Molecular Weight of Antigen 51.9 kDa
Purification Method Affinity Purified
Quantity 100 μg
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 1802
Reconstitution Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended.
Target Species Human, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.