Learn More
Description
Specifications
Specifications
| Antigen | DPH2 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | OTI4E7 |
| Conjugate | Biotin |
| Dilution | Western Blot, Flow Cytometry, Immunohistochemistry |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Alias | diphthamide biosynthesis protein 2, Diphthamide biosynthesis protein 2 homolog-like 2, diptheria toxin resistance protein required for diphthamide biosynthesis-like 2, diptheria toxin resistance protein required for diphthamide biosynthesis-like 2(S. cerevisiae), DPH2 homolog, DPH2 homolog (S. cerevisiae), DPH2L2diphthamide biosynthesis-like protein 2, DPH2-like 2, DPH2-like 2 (S. cerevisiae), HsDph2 |
| Gene Symbols | DPH2 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
