Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPM3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP226105 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP226105 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP226105 Supplier Novus Biologicals Supplier No. NBP226105
Add to Cart
Add to Cart

Goat Polyclonal Antibody

DPM3 Polyclonal specifically detects DPM3 in Human samples. It is validated for Western Blot, Peptide ELISA.

Specifications

Antigen DPM3
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:1000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Alias CDG1O, Dolichol-phosphate mannose synthase subunit 3, dolichol-phosphate mannosyltransferase subunit 3, Dolichyl-phosphate beta-D-mannosyltransferase subunit 3, dolichyl-phosphate mannosyltransferase polypeptide 3, DPM synthase complex subunit 3, Mannose-P-dolichol synthase subunit 3, MGC125904, MGC125905, MGC34275, MPD synthase subunit 3, prostin 1, prostin-1
Gene Symbols DPM3
Host Species Goat
Immunogen Peptide with sequence HDCEDAARELQSQ corresponding to internal region according to NP_061846.2, NP_714963.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54344
Test Specificity This antibody is expected to recognize both reported isoforms (NP_061846.2 and NP_714963.1).
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.