Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ DPO-1
GREENER_CHOICE

Catalog No. p-4783925
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Blocker of KV1.5 channel and IKur current

Blocker of IKur ultrarapid delayed rectifier potassium current and KV1.5 channels (IC50 = 0.31 μM for rKV1.5). Displays selectivity for inhibition of IKur over Ito (8-fold), IK1, IKr and IKs (20-fold) in native myocytes and selectivity for rat recombinant KV1.5 over KV3.1 (∽ 15-fold). Increases action potential duration in atrial but not ventricular myocytes and prevents atrial arrhythmia.

Chemical Identifiers

CAS 43077-30-1
Molecular Formula C22H29OP

Specifications

Quantity 10 mg
Formula Weight 340.44
Purity >99%
Molecular Weight (g/mol) 340.45
Chemical Name or Material DPO-1
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.