Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPP10 Antibody (004), PerCP, Novus Biologicals™
SDP

Catalog No. NB317210 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB317210 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB317210 Supplier Novus Biologicals Supplier No. NBP289978PCP
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

DPP10 Monoclonal antibody specifically detects DPP10 in Human samples. It is validated for ELISA

Specifications

Antigen DPP10
Applications ELISA
Classification Monoclonal
Clone 004
Conjugate PerCP
Dilution ELISA
Formulation PBS
Gene Alias dipeptidyl peptidase 10, Dipeptidyl peptidase IV-related protein 3, dipeptidyl peptidase like protein 2, Dipeptidyl peptidase X, Dipeptidyl peptidase-like protein 2, dipeptidyl-peptidase 10, dipeptidyl-peptidase 10 (non-functional), DPL2, DPP X, DPPY, DPRP-3, DPRP3dipeptidylpeptidase 10, inactive dipeptidyl peptidase 10, KIAA1492
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human DPP10 (Uniprot#: Q8N608-1; Leu 56-Glu 796).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57628
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.