Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPP3 Antibody (OTI2D9), Novus Biologicals™
SDP

Catalog No. NBP201007 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP201007 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP201007 Supplier Novus Biologicals Supplier No. NBP201007
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DPP3 Monoclonal specifically detects DPP3 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen DPP3
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI2D9
Concentration 0.58 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-2000, Flow Cytometry 1:100, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:150
Gene Alias Dipeptidyl aminopeptidase III, Dipeptidyl arylamidase III, dipeptidyl peptidase 3, Dipeptidyl peptidase III, dipeptidylpeptidase 3, dipeptidyl-peptidase 3, dipeptidylpeptidase III, DPP III, DPPIII, EC 3.4.14.4, FLJ11387, FLJ22331
Gene Symbols DPP3
Host Species Mouse
Immunogen Full length human recombinant protein of human DPP3(NP_005691) produced in HEK293 cell.
Molecular Weight of Antigen 82.4 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10072
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.