Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPP6 Antibody (274308), PE/Cy7, Novus Biologicals™
SDP

Catalog No. NB313541 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB313541 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB313541 Supplier Novus Biologicals Supplier No. FAB2360PECY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DPP6 Monoclonal specifically detects DPP6 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen DPP6
Applications Flow Cytometry
Classification Monoclonal
Clone 274308
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias dipeptidyl aminopeptidase IV-related protein, dipeptidyl aminopeptidase-like protein 6, Dipeptidyl aminopeptidase-related protein, Dipeptidyl peptidase 6, Dipeptidyl peptidase IV-like protein, dipeptidyl peptidase IV-related protein, Dipeptidyl peptidase VI, dipeptidylpeptidase 6, dipeptidyl-peptidase 6, dipeptidylpeptidase VI, DPP VI, DPPXFLJ55680, MGC46605, VF2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human DPP6, Thr56-Asp803 (Glu458Val), Accession # AAA35761
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1804
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.