Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPPA2 Antibody (OTI1G10), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. N272426A700 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N272426A700 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N272426A700 Supplier Novus Biologicals Supplier No. NBP272426AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DPPA2 Monoclonal specifically detects DPPA2 in Human, Rat, Canine samples. It is validated for Western Blot.

Specifications

Antigen DPPA2
Applications Western Blot
Classification Monoclonal
Clone OTI1G10
Conjugate Alexa Fluor 700
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CT100, developmental pluripotency associated 2, developmental pluripotency-associated protein 2, PESCRG1cancer/testis antigen 100, Pluripotent embryonic stem cell-related gene 1 protein
Gene Symbols DPPA2
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-175 of human DPPA2 (NP_620170) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Developmental Biology, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 151871.0
Target Species Human, Rat, Canine
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.