Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPPA2 Antibody (Z31P3C4*F11), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP250230IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250230IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250230IR Supplier Novus Biologicals Supplier No. NBP250230IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DPPA2 Monoclonal specifically detects DPPA2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen DPPA2
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone Z31P3C4*F11
Conjugate DyLight 755
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CT100, developmental pluripotency associated 2, developmental pluripotency-associated protein 2, PESCRG1cancer/testis antigen 100, Pluripotent embryonic stem cell-related gene 1 protein
Gene Symbols DPPA2
Host Species Mouse
Immunogen Peptide Sequence -KMMKRLMTVE
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Developmental Biology, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 151871
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.