Learn More
Invitrogen™ DPPA2 Polyclonal Antibody

Description
DPPA2 Polyclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | DPPA2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.2% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | DPPA2 |
| Gene Accession No. | Q9CWH0 |
| Gene Alias | 2410088E07Rik; C80932; cancer/testis antigen 100; CT100; D19Mgi18; developmental pluripotency associated 2; developmental pluripotency-associated 2; developmental pluripotency-associated protein 2; DPPA2; ECAT15-2; ECSA; embryonic stem cell (ESC) associated transcript 15-2; embryonic stem cell-associated transcript 15-2 protein; LOC683282; PESCRG1; Phsecrg1; Pluripotent embryonic stem cell-related gene 1 protein; similar to developmental pluripotency-associated 2 |
| Gene Symbols | DPPA2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.