Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ DPPI 1c hydrochloride
GREENER_CHOICE

Catalog No. 27831
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-831 1 mg
27-831-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 27-831 Supplier Tocris Bioscience™ Supplier No. 2783/1
Add to Cart
Add to Cart
This item is not returnable. View return policy

Dipeptidyl peptidase IV (DPP-IV) inhibitor

Inhibitor of dipeptidyl peptidase IV (DPP-IV) (IC50 = 104 nM) that displays selectivity over enzymes with DPP-like activity (IC50 > 30 μM). Increases plasma GLP-1 levels and improves glucose tolerance in diabetic mice following oral glucose challenge.

Chemical Identifiers

CAS 866396-70-5
Molecular Formula C14H21ClN4O2

Specifications

Quantity 1 mg
Formula Weight 312.8
Purity >98%
Molecular Weight (g/mol) 312.798
Chemical Name or Material DPPI 1c hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.