Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPPIV/CD26 Antibody (009), Alexa Fluor™ 532
SDP

Catalog No. NB076630 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB076630 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB076630 Supplier Novus Biologicals Supplier No. NBP290696AF532

Rabbit Monoclonal Antibody

DPPIV/CD26 Monoclonal specifically detects DPPIV/CD26 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen DPPIV/CD26
Applications Flow Cytometry
Classification Monoclonal
Clone 9
Conjugate Alexa Fluor 532
Formulation 50 mM Sodium Borate
Gene Alias ADABP, ADCP-2, ADCP2DPP IV, Adenosine deaminase complexing protein 2TP103, CD26 antigen, CD26T-cell activation antigen CD26, dipeptidyl peptidase 4, Dipeptidyl peptidase IV, dipeptidylpeptidase 4, dipeptidyl-peptidase 4, dipeptidylpeptidase IV (CD26, adenosine deaminase complexing protein 2), DPPIV, EC 3.4.14.5
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse DPPIV/CD26 (NP_034204.1; Ser29-His760).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cellular Markers, GPCR, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 1803
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.