Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPPIV/CD26 Antibody (202.36), HRP, Novus Biologicals™
SDP

Catalog No. NBP247256H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP247256H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP247256H Supplier Novus Biologicals Supplier No. NBP247256H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DPPIV/CD26 Monoclonal specifically detects DPPIV/CD26 in Human, Rat, Porcine (Negative), Sheep (Negative) samples. It is validated for Immunohistochemistry, Immunohistochemistry-Frozen.

Specifications

Antigen DPPIV/CD26
Applications Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 202.36
Conjugate HRP
Dilution Immunohistochemistry-Frozen
Formulation PBS with No Preservative
Gene Alias ADABP, ADCP-2, ADCP2DPP IV, Adenosine deaminase complexing protein 2TP103, CD26 antigen, CD26T-cell activation antigen CD26, dipeptidyl peptidase 4, Dipeptidyl peptidase IV, dipeptidylpeptidase 4, dipeptidyl-peptidase 4, dipeptidylpeptidase IV (CD26, adenosine deaminase complexing protein 2), DPPIV, EC 3.4.14.5
Gene Symbols DPP4
Host Species Mouse
Immunogen Human T cell clone
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cellular Markers, GPCR, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 1803
Test Specificity Recognizes a glycoprotein of 110kDa, identified as CD26 (Workshop VI; Code: N-L039). It is an atypical serine protease belonging to the prolyl oligopeptidase family. It is expressed on lymphocyte cells and is upregulated during T-cell activation. CD26 is also expressed on activated B cells and natural killer cells and abundantly on epithelia. CD26 is implicated in a variety of biological functions including T-cell activation, cell adhesion with extracellular matrix such as fibronectin or collagens, and in HIV infection. Cross-linking of CD26 using this antibody dramatically enhances the anti-CD3-induced IL-2 production. In Western blotting, this monoclonal antibody reacts with only glycosylated CD26, but not with the deglycosylated form. It does not prevent ADA binding to CD26.
Target Species Human, Rat, Porcine (Negative), Sheep (Negative)
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.