Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPPIV/CD26 Antibody (CACT114A), Novus Biologicals™
SDP

Catalog No. NBP260974 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP260974 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP260974 Supplier Novus Biologicals Supplier No. NBP260974

Mouse Monoclonal Antibody

DPPIV/CD26 Monoclonal specifically detects DPPIV/CD26 in Bovine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.

Specifications

Antigen DPPIV/CD26
Applications Flow Cytometry, Immunohistochemistry, Functional Assay
Classification Monoclonal
Clone CACT114A
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:10 - 1:1000
Formulation Ascites with 0.09% Sodium Azide
Gene Alias ADABP, ADCP-2, ADCP2DPP IV, Adenosine deaminase complexing protein 2TP103, CD26 antigen, CD26T-cell activation antigen CD26, dipeptidyl peptidase 4, Dipeptidyl peptidase IV, dipeptidylpeptidase 4, dipeptidyl-peptidase 4, dipeptidylpeptidase IV (CD26, adenosine deaminase complexing protein 2), DPPIV, EC 3.4.14.5
Gene Symbols DPP4
Host Species Mouse
Immunogen Bovine ConA stimulated mononuclear leukocytes
Purification Method Unpurified
Quantity 0.1 mg
Research Discipline Adaptive Immunity, Cellular Markers, GPCR, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 1803
Target Species Bovine
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.