Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPPIV/CD26 Antibody (H207-1082), Allophycocyanin
SDP

Catalog No. NB10062997J Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10062997J 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10062997J Supplier Novus Biologicals Supplier No. NB10062997APC

Rat Monoclonal Antibody

DPPIV/CD26 Monoclonal specifically detects DPPIV/CD26 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen DPPIV/CD26
Applications Flow Cytometry
Classification Monoclonal
Clone H207-1082
Conjugate APC
Dilution Flow Cytometry
Gene Alias ADABP, ADCP-2, ADCP2DPP IV, Adenosine deaminase complexing protein 2TP103, CD26 antigen, CD26T-cell activation antigen CD26, dipeptidyl peptidase 4, Dipeptidyl peptidase IV, dipeptidylpeptidase 4, dipeptidyl-peptidase 4, dipeptidylpeptidase IV (CD26, adenosine deaminase complexing protein 2), DPPIV, EC 3.4.14.5
Gene Symbols DPP4
Host Species Rat
Immunogen Made to Mouse CD26
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cellular Markers, GPCR, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 1803
Test Specificity NB100-62997 recognises mouse CD26, which is also known as thymocyte-activating molecule (THAM). Murine CD26 is a type II integral membrane dipeptidyl-peptidase that is expressed at the cell surface as a heterodimer, of approximately 220 kDa, and has a soluble form in the plasma. The CD26 molecule is a multifunctional glycoprotein that has ectopeptidase activity and is also reported to be involved in cell adhesion, signal transduction in T lymphocytes and HIV infection. CD26 is differentially expressed on developing thymocytes and is upregulated on activated T-cells but is not expressed on resting and thioglycollate stimulated macrophages. Expression of CD26 has also been reported on cells from a variety of non-haematopoietic tissues including liver, intestine and kidney.
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.