Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPYSL4 Antibody, Novus Biologicals™
SDP

Catalog No. NBP211055 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.05 mg
0.1 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP211055 0.1 mg
NB018831 0.025 mg
NBP21105505 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NBP211055 Supplier Novus Biologicals Supplier No. NBP211055
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DPYSL4 Polyclonal specifically detects DPYSL4 in Human, Mouse, Rat, Porcine, Feline, Primate samples. It is validated for Western Blot.

Specifications

Antigen DPYSL4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:450
Formulation Tris/Glycine buffer, pH 7.4-7.8, HEPES,BSA 0.5%, glycerol 30%. with 0.02% Sodium Azide
Gene Alias CRMP-3, CRMP3Collapsin response mediator protein 3, dihydropyrimidinase-like 4, dihydropyrimidinase-related protein 4, DRP-4UNC33-like phosphoprotein 4, ULIP4ULIP-4
Gene Symbols DPYSL4
Host Species Rabbit
Immunogen Synthetic peptides corresponding to amino acids form DRP4 protein. The peptide was covalently modified to achieve desired antigenicity before coupling to carrier protein.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 10570
Test Specificity Alternate Nomenclature:NP_006417 This antibody labels two major bands in PC-DRP4 of approximate MW of 84, 93 and a minor 114kDa and a smaller band of 35kDa, the identity of smaller protein is not known but could represent a proteolysis product
Target Species Human, Mouse, Rat, Pig, Feline, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.