Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPYSL4 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$243.50 - $670.50

Specifications

Antigen DPYSL4
Dilution Western Blot 1:450
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB018831
SDP
View Documents
Novus Biologicals
NBP2110550.025MG
0.025 mg
Each for $243.50
Only null left
Add to Cart
 
NBP21105505
SDP
View Documents
Novus Biologicals
NBP2110550.05MG
0.05 mg
Each for $402.00
Only null left
Add to Cart
 
NBP211055
SDP
View Documents
Novus Biologicals
NBP211055
0.1 mg
Each for $670.50
Only null left
Add to Cart
 
Description

Description

DPYSL4 Polyclonal specifically detects DPYSL4 in Human, Mouse, Rat, Porcine, Feline, Primate samples. It is validated for Western Blot.
Specifications

Specifications

DPYSL4
Western Blot
Unconjugated
Mouse, Rat, Pig, Primate, Human, Feline
10570
Synthetic peptides corresponding to amino acids form DRP4 protein. The peptide was covalently modified to achieve desired antigenicity before coupling to carrier protein.
Primary
Western Blot 1:450
Polyclonal
Rabbit
CRMP-3, CRMP3Collapsin response mediator protein 3, dihydropyrimidinase-like 4, dihydropyrimidinase-related protein 4, DRP-4UNC33-like phosphoprotein 4, ULIP4ULIP-4
DPYSL4
IgG
Alternate Nomenclature:NP_006417 This antibody labels two major bands in PC-DRP4 of approximate MW of 84, 93 and a minor 114kDa and a smaller band of 35kDa, the identity of smaller protein is not known but could represent a proteolysis product
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.