Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ DQP 1105
GREENER_CHOICESDP

Catalog No. 449150 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-915-0 50 mg
44-911-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 44-915-0 Supplier Tocris Bioscience™ Supplier No. 4491/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

GluN2C/GluN2D-selective NMDA antagonist

Noncompetitive NMDA receptor antagonist; displays over 50-fold selectivity for GluN2D- and GluN2C-containing receptors over GluN2B-, GluK2-, GluA1- and GluN2A-containing receptors (IC50 values are 2.7, 8.5, 121, 153, 198 and 206 μM, respectively). Reduces frequency of channel opening.

Please refer to IUPHAR Guide to Pharmacology for the most recent naming conventions.

Chemical Identifiers

CAS 380560-89-4
Molecular Formula C29H24BrN3O4

Specifications

Quantity 50 mg
Formula Weight 558.42
Purity >97%
Molecular Weight (g/mol) 558.432
Chemical Name or Material DQP 1105
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.