Learn More
Dräger™ Haz Mat Simultest Test Kit
Simultaneously detects multiple contaminants
Alert:
No other type of tube should be substituted for use with these kits.
Includes:
  • accuro™ pump
  • Pump spare parts kit
  • Air currents kit
  • 39 in. (1m) extension hose with adapter (includes scoring tool and manifold)
  • Simultaneous test set opener
  • Deluxe tube opener
  • Laminated instruction cards
  • Three each of Simultaneous Test Set† I (tests for hydrogen chloride, hydrogen cyanide, carbon monoxide, ammonia, and nitrous fumes)
  • Three each of Simultaneous Test Set† II (tests for sulfur dioxide, chlorine, hydrogen sulfide, carbon dioxide, and phosgene)
  • Four each of Simultaneous Test Set† III (tests for alcohols, aliphatic hydrocarbons, aromatics, chlorinated hydrocarbons, and ketones)
  • Hard-plastic carrying case
Products 1
Catalog Number Mfr. No. Includes Price Quantity & Availability  
Catalog Number Mfr. No. Includes Price Quantity & Availability  
19-500-221
Accessories View Documents
Dräger™
4056447
Without Pump N/A N/A N/A
Description
Description
Provides Hazmat teams with onsite information about levels of inorganic/organic gases and vapors encountered during fires and afterward during salvage and overhaul operations.
  • Easy to use—just score and remove tube tips, insert special five-tube holder into manifold, and connect manifold to pump
  • After 10 complete pump strokes, tubes are ready for evaluation
†Each Simultest Set consists of five detector tubes in a rubber holder—ready for use. Each test set can be completed in one minute or less.
Order Info
Individual test sets and adapter also sold separately.
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.