Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dräger™ Pac™ III Sensors

Catalog No. p-3888245 Shop All Draeger Safety, Inc. Products
Click to view available options
Components Measured:
Carbon Monoxide-HC (XS)
Diethylamine
Dimethyl Sulfide
Hydrogen Sulfide (XS)
Hydrogen Sulfide-HC (XS)
Nitric Oxide
Oxygen (XS)
Phosgene (COCL2)
Measurement Range:
0 to 100
0 to 1000
0 to 10000
0 to 25%
0 to 3
0 to 40
Warranty:
1 Year
3 Years
6 Months

Catalog No. 19-815-831

Supplier: Dräger™ 6809130

Only null left
Add to Cart
Add to Cart

Provide fast and accurate responses to hazardous gas levels

  • For use with Pac™ III single gas monitors

Specifications

Components Measured Oxygen (XS)
Humidity Range 10 to 95%
Measurement Range 0 to 25%
Pressure Range 700 to 1300mbar
Resolution 0.1%
Temperature Range (English) -40°F to +122°F
Temperature Range (Metric) -40° to +50°C
Warranty 3 Years
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.