Learn More
Dräger™ Short-Term Detector Tubes: Sulfur Dioxide
Detector tubes
$161.04 - $197.62
Specifications
Components Measured Sulfur Dioxide
Quantity 10
View More Specs
Products 5
Catalog Number Mfr. No. Measurement Range Tube Type Response Time Price Quantity & Availability  
Catalog Number Mfr. No. Measurement Range Tube Type Response Time Price Quantity & Availability  
17-986-71C
View Documents
Dräger™
8101531
50 to 8000 Sulfur dioxide 50/b Not Applicable
Pack of 10 for $188.44
Only null left
Add to Cart
17-985-301
View Documents
Dräger™
CH24201
20 to 2000 Sulfur dioxide 20/a 3 min.
Pack of 10 for $197.62
Only null left
Add to Cart
17-985-299
View Documents
Dräger™
6728491
0.5 to 25 Sulfur dioxide 0.5/a2 3 to 6 min.
Pack of 10 for $179.38
Only null left
Add to Cart
17-985-300
View Documents
Dräger™
CH31701
1 to 25 Sulfur dioxide 1/a 3 min.
Pack of 10 for $179.38
Only null left
Edge
Add to Cart
17-985-298
View Documents
Dräger™
6727101
0.1 to 3 Sulfur dioxide 0.1/a 20 min.
Pack of 10 for $161.04
Only null left
Add to Cart
Description
Description
Glass detector tubes are filled with an inert reagent carrier material (silica gel, aluminum oxide, or aluminum silicate) impregnated with an indicating reagent. Reagent produces a colorimetric indication in the presence of a particular gas, vapor, or aerosol. Concentrations can be read directly from the color change on the tube's printed scale. Tubes are hermetically sealed. Most have a two-year shelf life (when stored at room temperature). Available in packs of ten.
Note:
Some tests require a pretube or comparison tube in addition to the detector tube. These tubes are included in the box of 10, thus reducing the number of detector tubes. The actual number of detector tubes is cited in the ordering charts.
Order Info
Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
Specifications
Specifications
Sulfur Dioxide
10
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.