Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dräger™ X-act™ 5000 Automatic Tube Pump: Battery Packs

$15.29 - $766.44

Specifications

For Use With (Equipment) X-act 5000 Automatic Tube Pump
View More Specs

Products 3
Catalog Number Mfr. No. Type Voltage Price Quantity & Availability  
Catalog Number Mfr. No. Type Voltage Price Quantity & Availability  
19-453-022
View Documents
Dräger™
4523520
NiMH Battery Pack 110V
Each for $766.44
Only null left
Add to Cart
 
19-453-024
View Documents
Dräger™
4588195
AA Alkaline Battery Pack 1.5V
Pack for $15.29
Only null left
Add to Cart
 
19-453-023
View Documents
Dräger™
4523525
Alkaline Battery Pack 110V N/A N/A N/A
Description

Description

Specifications

Specifications

X-act 5000 Automatic Tube Pump
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.