Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DR3/TNFRSF25 Antibody (JD3) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB119738 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB119738 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB119738 Supplier Novus Biologicals Supplier No. NBP309115
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DR3/TNFRSF25 Monoclonal specifically detects DR3/TNFRSF25 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen DR3/TNFRSF25
Applications Flow Cytometry
Classification Monoclonal
Clone JD3
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Flow Cytometry 1-4 ug/ml
Formulation PBS
Gene Alias Apo-3, APO3, Apoptosis-inducing receptor AIR, Apoptosis-mediating receptor DR3, Apoptosis-mediating receptor TRAMP, Death receptor 3, DR3member 12 (translocatingchain-association membrane protein), Lymphocyte-associated receptor of death, Protein WSL, Protein WSL-1, tumor necrosis factor receptor superfamily, member 25, WSL, WSL1
Host Species Mouse
Immunogen human DR3/TNFRSF25-Ig fusion protein
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis, Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 8718
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.