Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DR3/TNFRSF25 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10056380SS
Description
DR3/TNFRSF25 Polyclonal specifically detects DR3/TNFRSF25 in Human samples. It is validated for Western Blot.Specifications
DR3/TNFRSF25 | |
Polyclonal | |
Unconjugated | |
PBS containing 0.2% gelatin with 0.05% Sodium Azide | |
Apo-3, APO3, Apoptosis-inducing receptor AIR, Apoptosis-mediating receptor DR3, Apoptosis-mediating receptor TRAMP, Death receptor 3, DR3member 12 (translocatingchain-association membrane protein), Lymphocyte-associated receptor of death, Protein WSL, Protein WSL-1, tumor necrosis factor receptor superfamily, member 25, WSL, WSL1 | |
Rabbit | |
Protein G purified | |
RUO | |
Primary | |
In HeLa, a 50 kDa band should be observed. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
0.5 mg/mL | |
Western Blot 0.5-2 ug/ml | |
NP_042997 | |
TNFRSF25 | |
The antibody was developed by immunizing rabbits with a synthetic peptide corresponding to amino acids 398-417 of human DR3 protein. | |
0.025 mg | |
Apoptosis, Cell Biology | |
8718 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction